Trimming knives
Toimittaja: SWANN MORTON
4206 4205 4204 4203 4202 4201 4313 4315
SWAN4206EA 99.5 EUR
SWAN4206 SWAN4205 SWAN4204 SWAN4203 SWAN4202 SWAN4201 SWAN4313 SWAN4315
Trimming knives
Veitset
Heavy duty trimming knifes with fixed or retractable blade.
  • Blades can be exchanged
  • Trimming knife with a smart black holster that clips onto a belt or waistband
Toimitustiedot: The fixed trimming knife is supplied with holster and three SM92 industrial blades. The retractable knife is supplied with two SM92 industrial blades.