Industrial knives, with exchangeable blades
Toimittaja: SWANN MORTON
4213 4212 4211 4214 4303 4302 4253 4252 4307
SWAN4213EA 42.1 EUR
SWAN4213 SWAN4212 SWAN4211 SWAN4214 SWAN4303 SWAN4302 SWAN4253 SWAN4252 SWAN4307
Industrial knives, with exchangeable blades
Veitset
Heavy-duty cast metal handles with either fixed blade or retractable blade mechanism. Ideal for industrial applications as well as for D.I.Y. enthusiasts.
  • Fixed handle can be opened with the aid of a screwdriver or small coin for blade changing
  • Retractable handle can be opened by hand for blade changing
  • Handles delivered without blades, set includes blades