TraceCERT®, Supelco® Chromate Standard for IC, CRM
Toimittaja: Merck
40121-100ML
SUPL40121-100MLEA 52.5 EUR
SUPL40121-100ML
TraceCERT®, Supelco® Chromate Standard for IC, CRM
Standardiaineet ja vertailumateriaalit Anionic Standards Chromate(VI) Standards
Single component solution suitable as ion chromatography standard.
Sertifikaatit: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34).