Superspher® RP8, HPLC Columns
Toimittaja: Avantor
SUPERRP84250AFEA 1150 EUR
SUPERRP84250AF SUPERRP8E4250F
Superspher® RP8, HPLC Columns
Kromatografiakolonnit
A range of 4 µm materials has been marketed under the Superspher® tradename in Europe.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
  • Hichrom high efficiency packed columns