Workstation Spilltray and Drying Rack
Toimittaja: Cole Parmer
67035-01
STUA67035-01EA 0 EUR
STUA67035-01
Workstation Spilltray and Drying Rack
Koeputkitelineet Kuivatustelineet
Reduce drying time and contain spilled liquids with this chemical resistant containment tray. Holds one ¹/₄ L of spilled liquid. Easy to clean simply lift off the white grid and empty.
  • Chemical resistant
  • Easy to clean
  • Dishwasher safe
  • Material: Polyethylene(PE)
  • Length: 419 mm
Tilaustiedot: One year warranty.
New Product