Cole-Parmer® Stuart SH-200 Microplate Shakers
Toimittaja: Cole Parmer
51901-08 51901-16
STUA51901-08EA 3030 EUR
STUA51901-08 STUA51901-16
Cole-Parmer® Stuart SH-200 Microplate Shakers
Ravistelijat Ravistelijat mikrotiitterilevyille
Provides a fast-mixing action in a tight orbit.
  • Non-slip mat and platform lip securely hold plates
  • Built-in timer and digital speed LED display provide reproducible results
  • Can be use in incubators
These microplate shakers combine high speed and a tiny 1.5 mm orbit to produce vigorous shaking which is perfect for microwell plates. The shakers offer a quiet drive mechanism, easy-to-use controller with timer, LED display, and a digital speed selection from 250 to 1250 rpm with a soft start.
Easily set digital timer from 1 to 999 minutes. When timerMore Product Information