Undergrad analogue magnetic hotplate stirrers, Stuart®
Toimittaja: Cole Parmer
04807-60 04807-58
STUA04807-60EA 770 EUR
STUA04807-60 STUA04807-58
Undergrad analogue magnetic hotplate stirrers, Stuart®
Sekoittajat Lämmittävät magneettisekoittajat
Get true temperature setting and accurate stirring control.
  • Glazed with BioCote® for antimicrobial protection
  • Compact, stackable design takes up less benchtop and storage space
  • Hot warning light remains on even when unplugged
  • Built with tough aluminium casing for longevity
  • Designed to prevent accidental spillage on control panel
  • IP 32 rated for water and dust resistance
Designed with microprocessor-controlled dual thermocouples, these stirring hot plates offer accurate temperature control and protection from overheating. The analogue models feature aMore Product Information