Cole-Parmer® Stuart INC-200 Clear Incubators
Toimittaja: Cole Parmer
01110-08 01110-02
STUA01110-08EA 5540 EUR
STUA01110-08 STUA01110-02
Cole-Parmer® Stuart INC-200 Clear Incubators
Inkubaattorit Pienikokoiset pöytämalliset inkubaattorit
Clear acrylic construction offers full visibility of samples.
  • Hinged front door provides easy access to working chamber
  • Each side panel features two 10 mm diameter ports for introducing gases or cables
Clear acrylic top and sides make it easy to view the sample from all sides - ideal for plant propagation, simulation of tropical conditions and corrosion testing. Forced air circulation and electronic temperature control ensure accurately maintained conditions. These incubators do not have a base so they can be placed directly over complete instruments;More Product Information