Human Recombinant Oncostatin M
Toimittaja: STEMCELL TECHNOLOGIES
78094 78094.1 78094.2
STMC78094EA 0 EUR
STMC78094 STMC78094.1 STMC78094.2
Human Recombinant Oncostatin M
Proteiinit
Oncostatin M (OSM) is a member of interleukin 6 (IL-6) family of cytokines and bears close resemblance to leukemia-inhibitory factor (LIF) and granulocyte colony-stimulating factor (G-CSF) in amino acid sequence and its modulation of differentiation in a variety of cell types (Rose and Bruce). OSM signals through type I receptor (consisting of gp130 and LIF receptor (LIFR)) and type II receptor (consisting of gp130 and OSM receptor (OSMR)), which eventually activate the JAK/STAT pathway (Auguste et al.; Gómez-Lechón). OSM is primarily produced by activated T cells and monocytes, and also by activated macrophages, neutrophils, mast cells, and dendritic cells. OSM is also produced within the bone microenvironment by cells of both hematopoietic and mesenchymal origin including osteocytes and osteoblasts. OSM is involved in differentiation, cell proliferation, hematopoiesis, and inflammation, and also has been shown to have implications in liver development, bone formation and resorption (Sims and Quinn; Tanaka and Miyajima).
Varoitus: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').