Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC)
Toimittaja: STEMCELL TECHNOLOGIES
60164FI 60164FI.1
STMC60164FIEA 0 EUR
STMC60164FI STMC60164FI.1
Anti-HLA-DR Mouse Monoclonal Antibody [clone: LN3] (FITC)
Vasta-aineet
Mouse monoclonal IgG2b antibody against human, rhesus HLA-DR, FITC-conjugated.
  • B cells
  • Dendritic cells
  • Macrophages
  • Monocytes
  • Other
  • T cells
The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II receptor encoded within the human leukocyte antigen (HLA) complex on chromosome 6. HLA-DR is a heterodimeric transmembrane glycoprotein comprising non-covalently associated α (36 kDa) and β (27 kDa) subunits. MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along withMore Product Information