Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (PE)
Toimittaja: STEMCELL TECHNOLOGIES
60022PE 60022PE.1
STMC60022PEEA 0 EUR
STMC60022PE STMC60022PE.1
Anti-CD8a Mouse Monoclonal Antibody [clone: RPA-T8] (PE)
Vasta-aineet
Mouse monoclonal IgG1 antibody against human, rhesus, cynomolgus CD8a, PE-conjugated.
  • T cells
  • T cells CD8
The RPA-T8 antibody reacts with CD8a, a 32 - 34 kDa type I transmembrane glycoprotein which is a subunit of CD8. CD8 is a disulfide-bonded dimer, found either as a heterodimer of CD8a (α) and CD8b (β) subunits (i.e., αβ) or a homodimer (αα). CD8 acts as a co-receptor to the T cell receptor (TCR) during T cell activation by binding MHC Class I molecules presented by an antigen-presenting cell. It functions to strengthen the association between theMore Product Information