Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (PE)
Toimittaja: STEMCELL TECHNOLOGIES
60016PE 60016PE.1
STMC60016PEEA 0 EUR
STMC60016PE STMC60016PE.1
Anti-CD4 Mouse Monoclonal Antibody [clone: OKT4] (PE)
Vasta-aineet
Rat monoclonal IgG2a antibody against mouse CD4, FITC-conjugated.
  • T cells
  • T cells CD4
  • T cells Regulatory
The RM4-5 antibody reacts with mouse CD4, an ~55 kDa single-chain type 1 transmembrane glycoprotein and member of the immunoglobulin (Ig) superfamily; CD4 contains four extracellular Ig-like domains. CD4 is expressed at relatively high levels by most thymocytes and a subpopulation of T cells (T helper/inducer cells), and at lower levels on dendritic cells. In the mouse, CD4 is not expressed by monocytes/macrophages. CD4 binds to a non-polymorphicMore Product Information