Anti-CD25 Rat Monoclonal Antibody [clone: 3C7] (FITC)
Toimittaja: STEMCELL TECHNOLOGIES
60010FI 60010FI.1
STMC60010FIEA 0 EUR
STMC60010FI STMC60010FI.1
Anti-CD25 Rat Monoclonal Antibody [clone: 3C7] (FITC)
Vasta-aineet
Rat monoclonal IgG2b antibody against mouse CD25, FITC-conjugated.
  • T cells
  • T cells Regulatory
The 3C7 antibody reacts with murine CD25 (interleukin‐2 receptor α chain; IL‐2Rα), an ~55 kDa type 1 transmembrane glycoprotein expressed on the surface of T and B cell progenitors and activated mature B and T cells. CD25 has been used as a marker to identify CD4+/FoxP3+ regulatory T cells in mice. Mutations in CD25 result in severe immunodeficiency, underscoring the importance of the roles the protein plays in the differentiation, activation andMore Product Information