EasySep™ Mouse CD4+ T Cell Isolation Kits
Toimittaja: STEMCELL TECHNOLOGIES
19852 19852RF
STMC19852EA 0 EUR
STMC19852 STMC19852RF
EasySep™ Mouse CD4+ T Cell Isolation Kits
Solujen eristysliuokset
Immunomagnetic isolation of untouched mouse CD4+ T cells
  • Fast and easy-to-use
  • Up to 96% purity
  • No columns required
  • Untouched, viable cells
The EasySep™ Mouse CD4+ T Cell Isolation Kit is designed to isolate CD4+ T cells from single-cell suspensions of splenocytes or other tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-CD4+ T cells and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. DesiredMore Product Information