EasySep™ Direct Human CTC Enrichment Kit
Toimittaja: STEMCELL TECHNOLOGIES
19657
STMC19657EA 0 EUR
STMC19657
EasySep™ Direct Human CTC Enrichment Kit
Solujen eristysliuokset
Immunomagnetic isolation of circulating tumor cells directly from whole blood
  • >99,9% RBC depletion without the need for density gradient centrifugation, sedimentation, or lysis
  • Up to 3 log depletion of normal hematopoietic cells
  • Fast, easy-to-use and column-free
  • Isolated cells are untouched
The EasySep™ Direct Human CTC Enrichment Kit can be used to enrich Circulating Tumor Cells (CTCs) directly from human whole blood by immunomagnetic negative selection without the need for either density gradient centrifugation or RBC lysis. Isolated cells areMore Product Information