EasySep™ Human Pan-Granulocyte Isolation Kits
Toimittaja: STEMCELL TECHNOLOGIES
19259 19259RF
STMC19259EA 0 EUR
STMC19259 STMC19259RF
EasySep™ Human Pan-Granulocyte Isolation Kits
Solujen eristysliuokset
Immunomagnetic isolation of untouched human pan-granulocytes
  • Fast, easy-to-use and column-free
  • Up to 99% purity
  • Untouched, viable cells
The EasySep™ Human Pan-Granulocyte Enrichment Kit is designed to isolate granulocytes (eosinophils, basophils and neutrophils) from normal peripheral whole blood by negative selection. Undesired cells are targeted for removal with Tetrameric Antibody Complexes recognising non-granulocytes and magnetic particles. The labelled cells are separated using an EasySep™ magnet without the use of columns. Desired cells areMore Product Information