EasySep™ Human Naïve B Cell Enrichment Kits
Toimittaja: STEMCELL TECHNOLOGIES
19254 19254RF
STMC19254EA 0 EUR
STMC19254 STMC19254RF
EasySep™ Human Naïve B Cell Enrichment Kits
Solujen eristysliuokset
Immunomagnetic isolation of untouched human naïve B cells
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Isolated cells are untouched
The EasySep™ Human Naïve B Cell Enrichment Kit is designed to isolate naïve B cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-naïve B cells and dextran-coated magnetic particles. The labelled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into aMore Product Information