EasySep™ Mouse CD8a Positive Selection Kit II
Toimittaja: STEMCELL TECHNOLOGIES
18953 18953RF
STMC18953EA 0 EUR
STMC18953 STMC18953RF
EasySep™ Mouse CD8a Positive Selection Kit II
Solujen eristysliuokset
Immunomagnetic positive selection of CD8a+ cells from mouse splenocytes or other tissues.
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
  • Isolated cells are not fluorochrome labelled
The EasySep™ Mouse CD8a Positive Selection Kit II is designed to isolate CD8a+ cells from single-cell suspensions of splenocytes or other tissues by positive selection. Desired cells are targeted with antibody complexes recognising CD8a and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. CellsMore Product Information