EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kits
Toimittaja: STEMCELL TECHNOLOGIES
17889 17889RF
STMC17889EA 0 EUR
STMC17889 STMC17889RF
EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kits
Solujen eristysliuokset
Column-free immunomagnetic positive selection of human CD8+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99,9% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD8 Positive Selection Kit is designed to isolate CD8+ cells from fresh whole blood or buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD8 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using anMore Product Information