EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kits
Toimittaja: STEMCELL TECHNOLOGIES
17885 17885RF
STMC17885EA 0 EUR
STMC17885 STMC17885RF
EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kits
Solujen eristysliuokset
Column-free immunomagnetic positive selection of human CD33+ cells for HLA analysis.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Whole Blood CD33 Positive Selection Kit is designed to isolate highly purified myeloid (including CD14+ and CD66b+) cells from fresh whole blood by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD33 and dextran-coated magnetic particles. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interestMore Product Information