EasySep™ HLA Chimerism BC CD56 Positive Selection Kits
Toimittaja: STEMCELL TECHNOLOGIES
17875 17875RF
STMC17875EA 0 EUR
STMC17875 STMC17875RF
EasySep™ HLA Chimerism BC CD56 Positive Selection Kits
Solujen eristysliuokset
Column-free immunomagnetic positive selection of human CD56+ cells for HLA analysis
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
The EasySep™ HLA Chimerism Buffy Coat CD56 Positive Selection Kit II is designed to isolate CD56+ cells from freshly prepared buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD56 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using anMore Product Information