EasySep™ Release Human CD3 Positive Selection Kits
Toimittaja: STEMCELL TECHNOLOGIES
17751 17751RF
STMC17751EA 0 EUR
STMC17751 STMC17751RF
EasySep™ Release Human CD3 Positive Selection Kits
Solujen eristysliuokset
Immunomagnetic positive selection of particle-free human CD3+ cells.
  • Highly purified human CD3+ cells isolated in less than 30 minutes
  • No-wash removal of EasySep™ Releasable RapidSpheres™
Isolate highly purified and magnetic particle-free CD3+ cells from peripheral blood mononuclear cells or washed leukapheresis samples in as little as 30 minutes.
Using immunomagnetic positive selection, CD3+ cells are labeled with antibodies and magnetic particles, then separated without columns using an EasySep™ magnet. Unwanted cells are poured off, whileMore Product Information