EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II
Toimittaja: STEMCELL TECHNOLOGIES
17699 17699RF
STMC17699EA 0 EUR
STMC17699 STMC17699RF
EasySep™ Human 'Do-It-Yourself' Positive Selection Kit II
Solujen eristysliuokset
Immunomagnetic positive selection of human cells with your own mouse IgG1 antibody.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human "Do-It-Yourself" Positive Selection Kit II is designed to select any human cell type of interest with your own mouse IgG1 monoclonal antibody. Cells that express the surface antigen that is recognised by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet. The cocktail also contains an antibody to human Fc receptor to minimiseMore Product Information