EasySep™ 'Do-It-Yourself' Positive Selection Kit II
Toimittaja: STEMCELL TECHNOLOGIES
17698 17698RF
STMC17698EA 0 EUR
STMC17698 STMC17698RF
EasySep™ 'Do-It-Yourself' Positive Selection Kit II
Solujen eristysliuokset
Immunomagnetic positive selection of any cells with your own mouse IgG1 antibody.
  • Fast and easy-to-use
  • No columns
The EasySep™ 'Do-It-Yourself' Selection Kit II is designed to select any cell type of interest with your own mouse IgG1 monoclonal antibody. Cells that express the surface antigen that is recognised by the mouse IgG1 monoclonal antibody are targeted for magnetic separation using EasySep™ Dextran RapidSpheres™ and the EasySep™ Magnet.
This product replaces the EasySep™ 'Do-It-Yourself' Selection Kit (STMC18098).
Varoitus: ProductMore Product Information