RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktails
Toimittaja: STEMCELL TECHNOLOGIES
15026 15066
STMC15026EA 0 EUR
STMC15026 STMC15066
RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktails
Solujen eristysliuokset
Immunodensity isolation of untouched stem/progenitor cells from cord blood.
  • Fast and easy-to-use
  • Requires no special equipment or training
  • Untouched, viable cells
  • Can be combined with SepMate™ for consistent, high-throughput sample processing
The RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail is designed to isolate progenitor cells from cord blood and whole blood by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD61, CD66b andMore Product Information