EasySep™ Human CD11b Positive Selection and Depletion Kits
Toimittaja: STEMCELL TECHNOLOGIES
100-0742 100-0744
STMC100-0742EA 0 EUR
STMC100-0742 STMC100-0744
EasySep™ Human CD11b Positive Selection and Depletion Kits
Solujen eristysliuokset
Immunomagnetic positive selection of CD11b+ cells from human peripheral blood mononuclear cells (PBMCs) or leukapheresis samples.
  • Fast and easy-to-use
  • Up to 99% purity
  • No columns required
Efficiently isolate or deplete human CD11b+ cells from fresh or previously frozen peripheral blood mononuclear cells in as little as 16 minutes. Cells isolated with the EasySep™ Human CD11b Positive Selection and Depletion Kit can be used immediately in downstream applications such as flow cytometry, cell culture, or DNA/RNA extraction.
Use this kit to label CD11b+More Product Information