EasySep™ Release Mouse CD138 Positive Selection Kits
Toimittaja: STEMCELL TECHNOLOGIES
100-0601 100-1440
STMC100-0601EA 0 EUR
STMC100-0601 STMC100-1440
EasySep™ Release Mouse CD138 Positive Selection Kits
Solujen eristysliuokset
Immunomagnetic positive selection kit using particle release technology.
  • Up to 95% purity of plasma cells from immunised mouse samples
  • No-wash removal of EasySep™ Releasable RapidSpheres™
  • Compatible with hybridoma generation protocols, including electrofusion
Isolate magnetic particle-free CD138+ plasma cells and plasmablasts from single-cell suspensions of mouse splenocytes or bone marrow in as little as 37 minutes. CD138+ cells are differentiated B cells that can be used in antibody discovery research due to their ability to generate affinity-More Product Information