STEMdiff™ Neural Progenitor Freezing Medium
Toimittaja: STEMCELL TECHNOLOGIES
05838
STMC05838EA 0 EUR
STMC05838
STEMdiff™ Neural Progenitor Freezing Medium
Solujen syväjäädytysliuokset
For cryopreservation of neural progenitor cells generated using STEMdiff™ Neural induction medium.
  • Serum-free
  • Optimised for cryopreservation of NPCs, with reproducibly high recovery rates
  • Supports cryopreservation of NPCs generated using STEMdiff™ Neural induction medium and cultured in STEMdiff™ Neural progenitor medium
  • Preserves NPC multipotency and expansion capacities
  • Convenient, user-friendly format and protocol
STEMdiff™ Neural Progenitor freezing medium is a serum-free medium for cryopreservation of neural progenitor cells (NPCs) derived from human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells. This freezing medium is optimised for the cryopreservation of NPCs generated using STEMdiff™ Neural induction medium and cultured in STEMdiff™ Neural progenitor medium. NPCs can be frozen at any point post neural induction, with reproducibly high recovery rates. Post thawing, NPCs display healthy morphology, express NPC markers, and retain the potential to expand and differentiate into neurons.
Varoitus: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').