Mouse IgA ELISA Antibody Pair Kit
Toimittaja: STEMCELL TECHNOLOGIES
01996
STMC01996EA 0 EUR
STMC01996
Mouse IgA ELISA Antibody Pair Kit
Bio-assayt ELISA-kitit
For detection and measurement of mouse immunoglobulin A.
The Mouse Immunoglobulin A (IgA) ELISA antibody pair kit is intended for those who want the flexibility of setting up their own ELISA assay. This kit includes capture and detection antibodies and an IgA standard.
It is designed for the quantitative detection and measurement of total mouse IgA in biological fluids such as serum, plasma, and cell culture supernatants. IgA is predominantly found on the surface of epithelial cells, where it acts as a neutralizing antibody against invading pathogens.
IgA primarily exists as a dimer in secretions such as saliva, tears, colostrum, mucus, sweat, and gastric fluid, but also occurs as a monomer in serum, where it comprises 10.
Varoitus: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').