Matrix, CELLvo™
Toimittaja: STEMBIOSYS
BM-HPME-6WP BM-HPME-T-75 BM-HPME-T-150
STBSBM-HPME-6WPEA 180 EUR
STBSBM-HPME-6WP STBSBM-HPME-T-75 STBSBM-HPME-T-150
Matrix, CELLvo™
Soluviljelyyn suplementit ja lisäaineet
CELLvo™ Matrix is a ready to use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.
  • Matrix made of polystyrene
  • Mesenchymal stem cells grown on CELLvo™ Matrix proliferate more rapidly
  • Mesenchymal stem cells grown on CELLvo™ Matrix maintain a smaller cell size
  • Mesenchymal stem cells grown on CELLvo™ Matrix exhibit a higher percentage of cells expressing SSEA-4
CELLvo™ Matrix is a natural extracellular matrix synthesised in vitro by bone marrow stromal cells and includes the biochemistry and architecture normally found in the bone marrow. It is a ready-to-use cell culture substrate which provides a native microenvironment that replicates the 3-dimensional 'home' in which stem cells naturally reside and proliferate.
CELLvo™ Matrix is a protein extracellular matrix that mimics the normal microenvironment of stem cells. This 3-D matrix is produced by young adult human bone marrow cells. The final product is a thin, off-white film on cell culture flasks, dishes, and substrates.
Lisävarusteiden tiedot: Accessories not included.
Toimitustiedot: CELLvo™ Matrix arrives at room temperature but should be refrigerated at 2 to 8°C upon arrival. Avoid extended exposure to light.
Pakkaus: CELLvo™ Matrix is produced under aseptic conditions and should be handled using aseptic technique.
Varoitus: CELLvo™ Matrix is not for human use. This product should not be used for diagnostic or therapeutic purposes.. Handle CELLvo™ related products in accordance with established bio-safety practices.