Hollow fibre filter modules, mPES membrane, MidiKros® TC
Toimittaja: Spectrum Laboratories
T04-E003-05-N T04-E030-05-N T04-E010-05-S T04-E010-05-N T04-E005-05-S T04-E003-05-S T04-E030-10-S T04-E300-10-S T04-E005-05-N T04-E300-05-S T04-E300-05-N T04-E100-10-S T04-E100-10-N T04-E100-05-S T04-E030-05-S T04-E030-10-N T04-E070-05-S T04-E070-05-N T04-E050-10-S T04-E050-10-N T04-E050-05-S T04-E050-05-N T04-E100-05-N T02-E100-05-S T02-E500-05-S T02-E500-05-N T02-E300-10-S T02-E300-10-N T02-E300-05-S T02-E300-05-N T02-E100-10-N T02-E100-05-N T02-E070-05-S T02-E070-05-N T02-E050-10-S T02-E100-10-S T02-E500-10-N T02-E500-10-S T04-E500-05-N T04-E300-10-N T06-E500-10-S T06-E050-05-N T06-E030-10-S T06-E050-10-S T04-E500-10-S T04-E500-10-N T04-E500-05-S T06-E100-10-N T06-E500-10-N T06-E500-05-S T06-E500-05-N T06-E300-10-S T06-E300-10-N T06-E300-05-S T06-E050-05-S T06-E100-10-S T06-E050-10-N T06-E100-05-S T06-E100-05-N T06-E070-05-S T06-E070-05-N T02-E050-05-N T06-E300-05-N T02-E050-10-N T06-E030-05-S T06-E030-10-N T06-E030-05-N T02-E010-05-N T02-E030-10-S T02-E030-10-N T02-E030-05-S T02-E030-05-N T02-E010-05-S T02-E005-05-S T02-E005-05-N T02-E003-05-S T02-E003-05-N T06-E003-05-S T06-E003-05-N T06-E005-05-S T06-E010-05-N T02-E050-05-S T06-E005-05-N T06-E010-05-S
SPECT04-E003-05-NEA 826 EUR
SPECT04-E003-05-N SPECT04-E030-05-N SPECT04-E010-05-S SPECT04-E010-05-N SPECT04-E005-05-S SPECT04-E003-05-S SPECT04-E030-10-S SPECT04-E300-10-S SPECT04-E005-05-N SPECT04-E300-05-S SPECT04-E300-05-N SPECT04-E100-10-S SPECT04-E100-10-N SPECT04-E100-05-S SPECT04-E030-05-S SPECT04-E030-10-N SPECT04-E070-05-S SPECT04-E070-05-N SPECT04-E050-10-S SPECT04-E050-10-N SPECT04-E050-05-S SPECT04-E050-05-N SPECT04-E100-05-N SPECT02-E100-05-S SPECT02-E500-05-S SPECT02-E500-05-N SPECT02-E300-10-S SPECT02-E300-10-N SPECT02-E300-05-S SPECT02-E300-05-N SPECT02-E100-10-N SPECT02-E100-05-N SPECT02-E070-05-S SPECT02-E070-05-N SPECT02-E050-10-S SPECT02-E100-10-S SPECT02-E500-10-N SPECT02-E500-10-S SPECT04-E500-05-N SPECT04-E300-10-N SPECT06-E500-10-S SPECT06-E050-05-N SPECT06-E030-10-S SPECT06-E050-10-S SPECT04-E500-10-S SPECT04-E500-10-N SPECT04-E500-05-S SPECT06-E100-10-N SPECT06-E500-10-N SPECT06-E500-05-S SPECT06-E500-05-N SPECT06-E300-10-S SPECT06-E300-10-N SPECT06-E300-05-S SPECT06-E050-05-S SPECT06-E100-10-S SPECT06-E050-10-N SPECT06-E100-05-S SPECT06-E100-05-N SPECT06-E070-05-S SPECT06-E070-05-N SPECT02-E050-05-N SPECT06-E300-05-N SPECT02-E050-10-N SPECT06-E030-05-S SPECT06-E030-10-N SPECT06-E030-05-N SPECT02-E010-05-N SPECT02-E030-10-S SPECT02-E030-10-N SPECT02-E030-05-S SPECT02-E030-05-N SPECT02-E010-05-S SPECT02-E005-05-S SPECT02-E005-05-N SPECT02-E003-05-S SPECT02-E003-05-N SPECT06-E003-05-S SPECT06-E003-05-N SPECT06-E005-05-S SPECT06-E010-05-N SPECT02-E050-05-S SPECT06-E005-05-N SPECT06-E010-05-S
Hollow fibre filter modules, mPES membrane, MidiKros® TC
Suodattimet Tangenttivirtausjärjestelmät
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have ¹/₂" TC/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information