Microfiltration filters, MiniKap®
Toimittaja: Spectrum Laboratories
MK2M204V6N
SPECMK2M204V6NEA 1090 EUR
SPECMK2M204V6N
Microfiltration filters, MiniKap®
Suodattimet Membraanisuodatinkapselit
These Point-of-use microfiltration filters are excellent for water and gas filtration systems. MiniKap® HF filter provides the filtration efficiency of the 0,2 µm DynaFibre® and air vent fibre in a device conveniently equipped with a ¼'' MNPT, Luer-Lok or hose barb inlet connection.