Peristaltic Pumps, KrosFlo®
Toimittaja: Spectrum Laboratories
ACR1-U2S-01N
SPECACR1-U2S-01NEA 8000 EUR
SPECACR1-U2S-01N
Peristaltic Pumps, KrosFlo®
Pumput Peristalttiset pumput
Spectrum's KrosFlo Research I (KRI) system combines a digital peristaltic pump and a multi-purpose lab stand to provide ultimate convenience and efficiency for experiments involving fluid circulation and transfers. The easy-to-operate digital KRI pump drives the fluid stream through flexible tubing (to be order separately) while the incorporated stand and included holders secure the tubing, filters, reservoirs and other components in place.
  • Portable compact set-up saves space
  • Accurate and reliable speed control
  • Locking digital keypad and display
  • Easy-to-