Mesh woven filters, PEEK, Spectra Mesh®
Toimittaja: Spectrum Laboratories
146804 146802 148883 148862 148863 148941 148882 148942
SPEC146804EA 957 EUR
SPEC146804 SPEC146802 SPEC148883 SPEC148862 SPEC148863 SPEC148941 SPEC148882 SPEC148942
Mesh woven filters, PEEK, Spectra Mesh®
Suodattimet Membraanisuodattimet
These woven filters are constructed of individual strands woven into a mesh screen, characterised by precise mesh openings, percent open area and mesh thickness. Available in five material types with different physical, chemical and thermal characteristics to select from depending on your application. Available in discs or square sheets.
Water affinity: Very hydrophobic
Strength/durability: Medium
Adsorption: Very low (inert)
Best resistance: Nearly all
pH resistance: 1 to 14
Thermal stability: up to 250 ºC
Sterilisation: Autoclavable