Membrane dialysis discs and sheets, Spectra/Por®
Toimittaja: Spectrum Laboratories
132476 132478 132480 132482 132486 132488 132677 132686 132723
SPEC132476EA 427 EUR
SPEC132476 SPEC132478 SPEC132480 SPEC132482 SPEC132486 SPEC132488 SPEC132677 SPEC132686 SPEC132723
Membrane dialysis discs and sheets, Spectra/Por®
Suodattimet Membraanisuodattimet
Pre-cut membrane discs are used with Spectra/Por® MacroDialyzers and Equilibrium Dialyzers.
  • Cut square membrane sheets can be used for a variety of dialysis equipment