Vitamin A palmitate USP

Toimittaja: Spectrum Chemical
Varoitus

V1159-1LT V1159-5KGBL
SPCMV1159-1LTEA 2980 EUR
SPCMV1159-1LT SPCMV1159-5KGBL
Vitamin A palmitate USP
Vitamin A palmitate

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Kaava: C₃₆H₆₀O₂
MDL numero: MFCD00019414
CAS-numero: 79-81-2
EINECS: 201-228-5

Order Now

Testitulosten erittely

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR