Magnesiumsitraatti, vedetön USP

Toimittaja: Spectrum Chemical

M2036-12KG M2036-2.5KG M2036-500GM
SPCMM2036-12KGEA 3840 EUR
SPCMM2036-12KG SPCMM2036-2.5KG SPCMM2036-500GM
Magnesiumsitraatti, vedetön USP
Magnesiumsitraatti

Magnesium citrate, anhydrous, USP is often used as a saline laxative but is also used as a food additive and dietary supplement.


Spectrum chemical USP products are manufactured, packaged and stored under current good manufacturing practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Sertifikaatit: Certified Halal.

Kaava: C₆H₈O₇·nMg
MDL numero: MFCD00036422
CAS-numero: 3344-18-1

Order Now

Testitulosten erittely

Assay (Mg) 14,5 - 16,4%
pH of 5% Suspension at 25 °C 5,0 - 9,0
Loss on Drying Max. 2,0%
Chloride (Cl) Max. 0,05%
Sulphate Max. 0,2%
Arsenic (As) 3 ppm
Heavy Metals 50 ppm
Iron (Fe) 200 ppm
Calcium (Dried Basis) Max. 1,0%
Identification Passes Test
Residual Solvents Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR