Metyyli-4-hydroksibentsoaatin natriumsuola ≥95.0% NF

Toimittaja: Spectrum Chemical

M1277-125GM M1277-500GM M1277-12KG M1277-2.5KG
SPCMM1277-125GMEA 204 EUR
SPCMM1277-125GM SPCMM1277-500GM SPCMM1277-12KG SPCMM1277-2.5KG
Metyyli-4-hydroksibentsoaatin natriumsuola ≥95.0% NF
Metyyli-4-hydroksibentsoaatin natriumsuola

Methylparaben Sodium, NF is an excipient used as a preservative. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Kaava: C₈H₇NaO₃
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016470
CAS-numero: 5026-62-0

Order Now

Testitulosten erittely

Assay (Anhydrous Basis) 95.0 - 102.0 %
Completeness of Solution To Pass Test
pH of a 1 in 1000 Solution 9.5 - 10.5
Water max. 5.0 %
Chloride max. 0.035 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR