Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910

Toimittaja: Spectrum Chemical

H1128-2.5KG H1128-500GM H1128-125GM
SPCMH1128-2.5KGEA 908 EUR
SPCMH1128-2.5KG SPCMH1128-500GM SPCMH1128-125GM
Hydroxypropyl methylcellulose (Hypromellose), 4000 mPa.s USP Substitution Type 2910
Hydroxypropylmethylcellulose

Hypromellose, Substitution Type 2910, 4000 mPa.s, USP is a semisynthetic, viscoelastic polymer that is inert and used as an ophthalmic lubricant, a controlled-delivery component and an excipient in oral medicaments. It is also used as a food additive and acts as an emulsifier, suspending agent and thickening agent. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Kaava: C₃H₈O₂·xCH₄O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00131360
CAS-numero: 9004-65-3

Order Now

Testitulosten erittely

Viscosity of a 2% Aqueous Solution @ 20D °C 3000 - 5600 mPa·s
Assay (Methoxy Groups) 28.0 - 30.0%
Assay:Hydroxypropoxy Groups 7.0 - 12.0%
pH 5.0 - 8.0
Loss On Drying max. 5.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR