Hydroxypropyl methylcellulose (Hypromellose), 50 cP FCC

Toimittaja: Spectrum Chemical

H1072-125GM H1072-2.5KG
SPCMH1072-125GMEA 169 EUR
SPCMH1072-125GM SPCMH1072-2.5KG
Hydroxypropyl methylcellulose (Hypromellose), 50 cP FCC
Hydroxypropylmethylcellulose

Hydroxypropyl Methylcellulose, 50 cps, FCC is most commonly used as an alternative to gelatin and gluten and is approved by FDA as both a direct and an indirect food additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Kaava: C₃H₈O₂·xCH₄O
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00131360
CAS-numero: 9004-65-3

Order Now

Testitulosten erittely

Assay (Methoxy Groups) 19.0 - 30.0%
Assay (Hydroxypropoxy Groups) 3.0 - 12.0 %
Viscosity of a 2% Aqueous Solution @ 20D °C 40 - 60 cPs
Lead (Pb) max. 3 mg/kg
Loss On Drying max. 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR