Rauta(III)oksidi ≥95%, jauhe, Reagent Grade

Toimittaja: Spectrum Chemical
Varoitus

F1035-2.5KG F1035-500GM F1035-50KG F1035-12KG
SPCMF1035-2.5KGEA 494 EUR
SPCMF1035-2.5KG SPCMF1035-500GM SPCMF1035-50KG SPCMF1035-12KG
Rauta(III)oksidi ≥95%, jauhe, Reagent Grade
Rauta(III)oksidi

Ferric Oxide, Powder, Reagent is an inorganic compound and is one of the main oxides of iron. It is used as a polishing agent (jeweler's rouge), a pigment and a photocatalyst. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Kaava: Fe₂O₃
Sulamispiste: 1538 °C
Tiheys: 5,24 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00011008
CAS-numero: 1309-37-1
EINECS: 215-168-2
Merck Index: 13,04058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR