Dextrin, white powder

Toimittaja: Spectrum Chemical

DE137-2.5KG DE137-500GM
SPCMDE137-2.5KGEA 275 EUR
SPCMDE137-2.5KG SPCMDE137-500GM
Dextrin, white powder
Dekstriini

Dextrin, White Powder is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Kaava: (C₆H₁₂O₆)n
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00081554
CAS-numero: 9004-53-9
EINECS: 232-675-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR