Diethylenetriamine ≥98%

Toimittaja: Spectrum Chemical

D1092-200LTBL D1092-20LT
SPCMD1092-200LTBLEA 7370 EUR
SPCMD1092-200LTBL SPCMD1092-20LT
Diethylenetriamine ≥98%
Dietyleenitriamiini

Diethylenetriamine is also known under the abbreviation DETA and is a colorless liquid that is hygroscopic and soluble in both water and polar organic solvents. Used as a ligand forming complex in coordination chemistry, it is also used for the extraction of gases that are acid in the oil industry.


GHS hazard : Corrosion;Exclamation mark;Health hazard
UN2079

Kaava: C₄H₁₃N₃
Tiheys: 0,955
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00008171
CAS-numero: 111-40-0
UN: 2079
ADR: 8,II

Order Now

Testitulosten erittely

Assay 98% min.
Water max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR