Roottorit, Sorvall™ LYNX -sentrifugeihin
Toimittaja: Thermo Fisher Scientific
096-062185 096-061075 096-062075 096-062375 096-084275 75003000 AVL096-145075 096-124375 75003007 75003008 AVL096-062075 096-041075 75003010 75003002
521-1966EA 17733 EUR
521-1966 521-1964 521-1965 521-1967 SORV096-084275 521-1977 521-1968 SORV096-124375 SORV75003007 SORV75003008 SORVAVL096-062075 521-2670 SORV75003010 SORV75003002
Roottorit, Sorvall™ LYNX -sentrifugeihin
Sentrifugit Roottorit
BIOFlex™ HC Swinging-Bucket Rotor Process more than 35 tube styles from microtubes to 1L bottles, plus conical tubes, microplates, tissue culture flasks and blood bags with the Thermo Scientific™ BIOFlex™ HC Swinging-Bucket Rotor. Innovations such as Auto-Lock rotor exchange, Auto-ID instant rotor identification and speed handles on each rotor lid shorten run set-up time while providing peace of mind that the rotor is secure.
  • Secure, push-button rotor exchange in less than three seconds
  • Improved safety and convenience