Centrifuge bottles, PPCO/PC, Fiberlite™, Sorvall™
Toimittaja: SORVALL
010-1493 010-1494 010-1492 010-1495 010-1496 010-1491
SORV010-1493EA 333 EUR
SORV010-1493 SORV010-1494 521-2669 SORV010-1495 SORV010-1496 521-2668
Centrifuge bottles, PPCO/PC, Fiberlite™, Sorvall™
Pullot Sentrifugipullot
These Fiberlite™ bottle systems combine a Nalgene bottle body with a newly designed Fiberlite cap closure.
For use with Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A large-capacity centrifuges.