Functional model of the shoulder
Toimittaja: SOMMER MARCUS
NS53
SOMMNS53EA 0 EUR
SOMMNS53
Functional model of the shoulder
Mallit Ihmisbiologia
The following movements are possible:
  • Raising of the arm, abduction
  • Swinging of the arm, anteversion
  • Back swinging of the arm, retroversion
  • Inner and outer rotation
Removable from stand with green base.