Ear section through the cochlea
Toimittaja: SOMMER MARCUS
DS10
SOMMDS10EA 0 EUR
SOMMDS10
Ear section through the cochlea
Mallit Ihmisbiologia
The scala of the vestibule, the scala of the tympanum, the cochlear duct with tectorial membrane and organ of Corti are shown.
  • Enlarged approximately 350 times
  • On a green base plate