TriLink® 2',3'-Dideoxycytidine-5'-O-(1-Thiotriphosphate)
Toimittaja: TriLink BioTechnologies
N-8010-1 N-8010-10 N-8010-5
SOLUN-8010-1EA 94.3 EUR
SOLUN-8010-1 SOLUN-8010-10 SOLUN-8010-5
TriLink® 2',3'-Dideoxycytidine-5'-O-(1-Thiotriphosphate)
Nukleiinihapporeagenssit Nukleotidit
2',3'-Dideoxycytidine-5'-O-(1-Thiotriphosphate) is a sugar modified nucleoside triphosphate where the 2' and 3' hydroxyl groups are absent, resulting in chain termination. This ddCTP analog also contains a 1-Thiotriphosphate which results in a nuclease resistant phosphorothioate linkage. The inability of polymerases to extend from a dideoxy nucleotide causes chain termination and is useful in antiviral research and in a variety of biotechnology applications.
  • Grade: RUO
Varoitus: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a62e67e40193d29cef8952559db39211c7e3d56&vid=3a62e67e40193d29cef8952559db39211c7e3d56&prid=web&clid=SAPDAM