TriLink® Adenosine-5'-Triphosphate
Toimittaja: TriLink BioTechnologies
N-1501-10 N-1501-100 N-1501-25
SOLUN-1501-10EA 25.3 EUR
SOLUN-1501-10 SOLUN-1501-100 SOLUN-1501-25
TriLink® Adenosine-5'-Triphosphate
Nukleiinihapporeagenssit Nukleotidit
This nucleotide consists of an unmodified base, a ribose, and a 5′ triphosphate group for synthesis of RNA molecules.
  • Unmodified ribonucleotide for RNA synthesis
  • >99% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
Varoitus: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38af3e784fe47c380861c8d1226b583e7c239779&vid=38af3e784fe47c380861c8d1226b583e7c239779&prid=web&clid=SAPDAM