TriLink® 5-Methoxyuridine-5'-Triphosphate
Toimittaja: TriLink BioTechnologies
N-1093-1 N-1093-10 N-1093-100 N-1093-5
SOLUN-1093-1EA 75.3 EUR
SOLUN-1093-1 SOLUN-1093-10 SOLUN-1093-100 SOLUN-1093-5
TriLink® 5-Methoxyuridine-5'-Triphosphate
Nukleiinihapporeagenssit Nukleotidit
This nucleotide consists of a modified base, a ribose, and a 5′ triphosphate group.
  • Can be incorporated in in vitro transcription for modification of synthesized RNA for desired characteristics
  • ≥97% purity by AX-HPLC analysis, provided as 100 mM in H₂O
  • Grade: RUO
5-Methoxyuridine-5'-Triphosphate is a modified NTP for incorporation into messenger RNAs (mRNA) by in vitro transcription using T7 RNA Polymerase. Incorporation of 5-Methoxyuridine can reduce the immunostimulatory effects of the resulting mRNAs.
Varoitus: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c04fde5b67579788608646f5bb1d814c5f2d59&vid=95c04fde5b67579788608646f5bb1d814c5f2d59&prid=web&clid=SAPDAM